Learn More
Cytiva Whatman™ Glass Microfiber Extraction Thimbles, Grade 603G
Product Code. 11393105
Change view
Click to view available options
Diameter (Metric) Inside:
10 mm
16 mm
19 mm
22 mm
23.8 mm
25 mm
28 mm
30 mm
33 mm
35 mm
Length (Metric):
38 mm
50 mm
60 mm
68 mm
80 mm
90 mm
94 mm
100 mm
118 mm
150 mm
Wall Thickness:
1 mm
1.5 mm
Unit Size:
Pack of 25
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Length (Metric) Wall Thickness Unit Size
11393105 35 mm 150 mm 1.5 mm Pack of 25
11363095 16 mm 50 mm 1 mm Pack of 25
11303105 25 mm 100 mm 1.5 mm Pack of 25
11373095 19 mm 90 mm 1 mm Pack of 25
11383095 22 mm 80 mm 1.5 mm Pack of 25
11353105 30 mm 100 mm 1.5 mm Pack of 25
11343105 28 mm 60 mm 1.5 mm Pack of 25
11383105 33 mm 94 mm 1.5 mm Pack of 25
11363105 33 mm 100 mm 1.5 mm Pack of 25
11393095 23.8 mm 68 mm 1.5 mm Pack of 25
11373105 33 mm 118 mm 1.5 mm Pack of 25
11353095 10 mm 38 mm 1 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11393105 Supplier Cytiva Supplier No. 10371055
  • ONLINE EXCLUSIVE PRICE
    358.00€
    344.00€ / Pack of 25
    Save 14.00€
    3% Off
View all promotions
In Stock
Flexibly designed for use in a full range of commercially available Soxhlet apparatus systems. Whatman™ Glass Microfiber Extraction Thimbles are designed for applications including analysis of hot and acidic gases, analysis of pesticide residues, determination of oil/fat content of foods, and analysis of oil and grease in solid wastes.
  • Resistant to temperatures up to 500°C (932°F)
  • Manufactured with walls as thin as 1–1.5mm
  • Standard glass Soxhlet thimble applications
  • 603G borosilicate glass fiber thimble with inorganic binder
TRUSTED_SUSTAINABILITY
Specifications
Description GE’s Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available Soxhlet apparatus systems.
Diameter (English) Inside 1.37 in.
Diameter (Metric) Inside 35 mm
Material Borosilicate Glass Fibers
Wall Thickness 1.5 mm
For Use With (Application) Smoke stack gas monitoring. Soxhlet extraction. Analyzing pesticide residues. Determining oil/fat content of foods. Analysis of oil and grease in solid wastes.
Grade 603G
For Use With (Equipment) Soxhlet extractor
Length (English) 5.9 in.
Length (Metric) 150 mm
Max. Temperature (English) 932°F
Max. Temperature (Metric) 500°C
Particle Retention 0.8μm
Product Type Extraction Thimble