Learn More
Cytiva Foil-Barrier Sample Bags
Product Code. 11984265
Change view
Click to view available options
Closure Type:
Zip Seal
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type Unit Size
11984265 Zip Seal Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11984265 Supplier Cytiva Supplier No. 10534321
  • ONLINE EXCLUSIVE PRICE
    83.50€
    78.69€ / Pack of 100
    Save 4.81€
    5% Off
In Stock
For use with 903 Protein Saver cards
  • Zip to lock
  • Resealable
  • For cards 95 x 130mm
  • For use with 903 protein saver cards
TRUSTED_SUSTAINABILITY
Specifications
Description Resealable
Closure Type Zip Seal
For Use With (Equipment) 903 Protein Saver cards, 95 x 130 mm
Product Type Sample Bag