missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Cyclopore PC Polycarbonate Membrane Filters

Microporous membranes featuring sharp cut-off and reproducible lab microfiltration characteristics of track-etched membrane filters

Brand:  Cytiva 10418550

Additional Details : Weight : 0.08000kg

 View more versions of this product

Product Code. 16655675

  • 176.00€ / Pack of 100
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • Low affinity for stains improves visibility under a microscope
  • Low nonspecific protein binding minimizes sample loss of proteinaceous samples
  • Narrow micropore size distribution and high pore density
    • Support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate
    • Nonhygroscopic
  • No particle shedding enables clean filtrate
  • Biologically inert

Applications:

  • General filtration
  • Cross-flow filtration (tangential flow filtration)
  • Air and water monitoring
  • Microorganism analysis
  • Healthcare applications
    • Immunodiagnostics
Specifications

Specifications

Autoclavable
12 μm
Min: 105 pores/cm2, Max: 6 x 108 pores/cm2
100 Pk.
140 °C
1.85 in.
Polycarbonate
For general filtration, Cross-flow filtration, Air and water monitoring, Microorganism analysis, Healthcare applications, Immunodiagnostics
Min: 4%, Max: 20%
284 °F
Membrane filters
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers