Learn More
Cytiva Whatman™ AS 300 Series Filtration Manifolds
Click to view available options
:
Filter funnel; 3-place manifold
Description
The stainless steel manifold for three or six filtration units is fitted with stainless steel units. The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
Specifications
Specifications
| Autoclavable | Autoclavable |
| For Use With (Equipment) | Microbiology monitors and analytical funnels |
| Material | Stainless Steel |
| Manifold Type | 3 Stopcocks and lower parts for individual choice of filter units |
| No. of Positions | 3 |
| Port Detail | 9 mm I.D. |