Learn More
Cytiva Whatman™ 1 Chr Chromatography Paper
Product Code. 11388804
Change view
Click to view available options
Length (Metric):
20 cm
25 cm
57 cm
100 m
Width (Metric):
2 cm
3.8 cm
4 cm
20 cm
25 cm
46 cm
Format:
Roll
Sheet
Unit Size:
Each
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Format unitSize
11388804 100 m 3.8 cm Roll Each
11390724 20 cm 20 cm Sheet Pack of 100
10387291 100 m 4 cm Roll Each
11310734 57 cm 46 cm Sheet Pack of 100
11320724 100 m 2 cm Roll Each
11300734 25 cm 25 cm Sheet Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11388804 Supplier Cytiva Supplier No. 3001651
Add to basket
Edge
Add to basket
Ensures the wicking capability and uniformity of capillary action important in chemical separations. Whatman™ 1 Chr Chromatography Paper is also widely used in protein and nucleic acid blotting.
Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind. Manufactured and tested specifically for chromatographic techniques to ensure the wicking capability and uniformity of capillary action important in chemical separations.
  • Typical Thickness: 0.18mm
  • Linear flowrate: 130mm/30 min.
  • Smooth surface
  • For general analytical separations
TRUSTED_SUSTAINABILITY
Specifications
Description 1 Chr cellulose chromatography roll is a 0.18 mm paper for general analytical separations.
Dimensions (L x W) 38.1 cm x 91.44 m (1.5 in. x 300 ft.)
Film Thickness 0.18 mm
Finish Smooth
For Use With (Application) For General Analytical Separations
Format Roll
Grade Grade 1 Chr
Material Cellulose
Width (English) 1.5 in.
Width (Metric) 3.8 cm
Flow Rate 130 mm/30 min.
Length (Metric) 100 m