Learn More
Thermo Scientific™ Well Plate Accessories
Product Code. 15102809
Click to view available options
Product Type:
Automated Sample Concentrator
Center Hole Cap
Deep Well Plate Vial
MTRS Vial
Manual Concentrator
Mat Applicator
Microtiter Plate Vial
Plug
Sealing Tape Applicator
Square Well Plate Vial
Tube Tray
Volume (Metric) Working:
100 μL
1000 μL
1100 μL
1500 μL
350 μL
500 μL
700 μL
800 μL
Not Applicable
Well Shape:
Flat Bottom (F-well)
Not Applicable
Round
Square
Taper
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 2
Pack of 500
Pack of 96
Pack of 960
Product Code. 15102809
Brand: Thermo Scientific™ 60180C101
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Effectively seal all microplate formats to minimize evaporation and protect samples from contamination and spilling, ensure a consistent, reliable seal with all microplate formats with this easy-to-use, handheld applicator, and get significantly quicker dry-down times than standard methods, and remove most chromatography solvents in just minutes with easy to use 96-well sample concentrators.
Sealing Tape Applicator
Ensure a consistent, reliable seal with all microplate formats and all types of tape with this easy to use, handheld applicator for Thermo Scientific™ WebSeal Sealing Tapes. They are packaged two per box.
Sample Concentrators
Remove most chromatography solvents in just minutes. Simple to operate and maintain, Thermo Scientific™ WebSeal Sample Concentrators allow quick, easy solvent evaporation from 96 well microplates, offering significantly quicker dry-down times than standard methods such as vacuum ovens or centrifuges. These evaporators offer significant throughput advantages for laboratories looking to accelerate microplate sample preparation. Depending on your throughput requirements, choose either the manually operated MiniVap Concentrator or the programmable, robot-ready UltraVap.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Webseal
Color Blue
Product Type Plug
For Use With (Equipment) Micro-Tube-Rack System, 8 mm
Material Polyethylene
No. of Wells 96
Volume (Metric) Working Not Applicable
Well Shape Not Applicable
Includes PE Plug
Plate Compatibility 96-well Plates