Learn More
Thermo Scientific™ WebSeal™ 96-Well Mats for WebSeal Glass Inserted Plate Kits
Product Code. 15152789
Click to view available options
Pre-slit:
No
Yes
Product Type:
WebSeal Mat, Solid Plugs
WebSeal Mat, Welled Plugs
Unit Size:
Each
Product Code. 15152789
Brand: Thermo Scientific™ 60180M157
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Extend the usefulness of your well plate kits with replacement mats.
Extend the usefulness of your wellplate kits with WebSeal 96-Well Mats, which serve as accessories for WebSeal Glass Inserted Plate Kits.
Extend the usefulness of your wellplate kits with WebSeal 96-Well Mats, which serve as accessories for WebSeal Glass Inserted Plate Kits.
TRUSTED_SUSTAINABILITY
Specifications
Material Silicone, PTFE
No. of Wells 96
No. per Pack 1 Mat
For Use With (Application) Sample Storage
For Use With (Equipment) WebSeal Glass Inserted Plate Kit (Cat. No. 60180-K109)
Pre-slit No
Product Type WebSeal Mat, Solid Plugs
Product Line Webseal
Plate Compatibility WebSeal Glass Inserted 96-well Plate