Learn More
Thermo Scientific™ Webseal 384-Well Plate
384-Well Plate with square v-shaped base.
Brand: Thermo Scientific™ 60180-P109B
58.70 EUR valid until 2024-05-31
Use promo code "21712" to get your promotional price.
Additional Details : Weight : 0.20000kg
Description
- 384-well
- Polypropylene
- Square V Base
- Barcoded
Specifications
Without Lid | |
Webseal | |
Yes | |
Polypropylene | |
384 | |
Conical Bottom (V-well) |
Autoclavable | |
Clear | |
Full | |
145 μL | |
Non-sterile |