Learn More
Watson-Marlow Manifold Tubing - Marprene
Description
- Made from autoclavable marprene
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Specifications
Specifications
| Diameter (English) Inner | 3/50 in. |
| Diameter (Metric) Inner | 1.65 mm |
| Material | Marprene |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.8 mm |
| For Use With | 323S/MC peristaltic pump |
| Packaging Quantity | 15m/Pk. |
| Flow Rate | 0.27 to 19 (four roller pump), 0.2 to 14 (eight roller pump)mL/min. |
| Color | Blue/Blue |
| Product Type | Double Segment Tubing |