Learn More
Water WFI Quality, Corning™ Mediatech™
BIOPHARMA
Product Code. 15373651
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15373651 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15373651 Supplier Corning Media Tech Supplier No. 25055CVC
  • ONLINE EXCLUSIVE PRICE
    104.00€
    101.55€ / Pack of 6
    Save 2.45€
    2% Off
View all promotions
 Restricted Item
In Stock
Corning™ High Quality Water is manufactured in an FDA-registered facility in accordance with a comprehensive quality system compliant with cGMP and current ISO 13485 guidelines. Corning offers a range of high quality water products for small and large volume requirements. These products eliminate the expense of installing and maintaining your own water system, and ensure you have a secondary source of water that meets your specifications. Corning High-Quality water is available in a variety of pack sizes, and tested against specifications which may include USP and EP monograph requirements.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 mL