Learn More
Miele Professional™ Water Purification SK/1
Product Code. 30533242 Shop All Miele Products
Change view
Click to view available options
Includes:
VA plug connection
Unit Size:
1 sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
30533242 VA plug connection 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30533242 Supplier Miele Professional™ Supplier No. 12684790
For simple and convenient changing of demineralisation cartridges
  • 2 x VA plug connection 3/4, 2 x quick connection 3/4
  • 2 x VA plug connection 3/4 for installation on second cartridge
  • For simple and convenient changing of demineralisation cartridge
TRUSTED_SUSTAINABILITY
Specifications
Product Type Water Connection
For Use With (Equipment) Washer-Disinfectors
Includes VA plug connection
Quantity 1 Ea.