Learn More
Rotronic™ Water Activity Station Probe
Product Code. 13558550
Change view
Click to view available options
Material:
Stainless Steel, Aluminum
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
13558550 Stainless Steel, Aluminum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13558550 Supplier Rotronic™ Supplier No. HC2AW
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Accuracy ± 0.008 aw / 0.8 %rh / ±0.1 K
Material Stainless Steel, Aluminum
Temperature Range (Metric) -40°C to +85°C, 0 to 100% RH