Learn More
Agilent BioTek Waste Tubing for 50 TS Microplate Washer
GREENER_CHOICE
Product Code. 10186272 Shop All Agilent BioTek Products
Click to view available options
Unit Size:
Each
Product Code. 10186272
Brand: Agilent BioTek 4070511
Accessory for 50 TS microplate washer
  • Includes: rubber stopper
  • Length: 1.2m (4 ft.)
TRUSTED_SUSTAINABILITY
Specifications
Includes Rubber stopper
Type Waste Tubing Set