Learn More
Vitlab™ Wash Bottle Vent Cap
Click to view available options
Closure Size:
GL 45
GL 63
Color:
Red
Yellow
Description
- Screw cap and spray tube
- Practically no leakage or dripping
- Design prevents almost all static overpressure
- Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
Specifications
Closure Size | GL 45 |
Closure Type | Screw Cap |
Color | Yellow |
Description | Screw cap with spray tube |
Material | Polypropylene |