Learn More
(±)-Warfarin Sodium, TRC
Specifications
Specifications
| Chemical Name or Material | Warfarin sodium |
| CAS | 129-06-6 |
| Molecular Formula | C19 H15 O4 . Na |
| Quantity | 5 g |
| Synonym | Warfarin sodium, Sodium 2-oxo-3-[(1RS)-3-oxo-1-phenylbutyl]-2H-1-benzopyran-4-olate, 2H-1-Benzopyran-2-one','4-hydroxy-3-(3-oxo-1-phenylbutyl)-','sodium salt (1:1), 2H-1-Benzopyran-2-one','4-hydroxy-3-(3-oxo-1-phenylbutyl)-','sodium salt (9CI), Coumarin','3-(α-acetonylbenzyl)-4-hydroxy-','sodium salt (8CI), Warfarin','sodium deriv. (6CI), (±)-Warfarin sodium, 3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium, Aldocumar, Athrombin, Coumadan Sodico, Coumadin, Coumadin sodium, Coumadine, Coumafene sodium, Dimantil, Farin, Marevam, Marevan, Orfarin, Panwarfin, Prothromadin, Ratsul Soluble, Simarc 2, Sodium coumadin, Sodium warfarin, Sodium','[[2-oxo-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-4-yl]oxy]-, Sofarin, Taro-warfarin, Tintorane, UniWarfin, Varfine, Waran, Warfarin sodium, Warfarin sodium salt, Warfarina, Warfil 5, Warfilone, Zoocoumarin sodium salt |
| InChI Formula | InChI=1S/C19H16O4.Na/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22;/h2-10,15,21H,11H2,1H3;/q;+1/p-1 |
| SMILES | [Na+].CC(=O)CC(C1=C([O-])c2ccccc2OC1=O)c3ccccc3 |
| IUPAC Name | sodium;2-oxo-3-(3-oxo-1-phenylbutyl)chromen-4-olate |
| Molecular Weight (g/mol) | 330.31 |
| Formula Weight | 330.0868 |