Learn More
R&L Enterprises™ Ward Pattern Furnace Tongs
Product Code. 11869701 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
590 mm
740 mm
Unit Size:
Each
Product Code. 11869701
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For protection and safety when handling heated objects in the laboratory. R&L Enterprises™ Ward Pattern Furnace Tongs are fitted with a circular shield to protect the hand against radiation from the furnace.
  • Manufactured from stainless steel
  • For handling a wide variety of crucibles, especially in muffle furnaces
  • Operated by thumb action with one hand
Specifications
For Use With (Application) For handling a wide variety of crucibles and to protect the hand against radiation from the furnace
Quantity 1 Ea.
Length (Metric) 740 mm
Material Stainless Steel
Type Furnace Tongs