Learn More
R&D Systems™ VVD 844
Description
VVD 844 is a potent inhibitor of the PI3K/p110α interaction (IC50 = 4 nM) and the phosphorylated-Ser473-AKT (pAKT) pathway (IC50 = 11 nM). It covalently binds cysteine 242 in the RAS domain of PI3K/p110α and blocks RAS activation of PI3Kα activity in vitro.
In vivo, VVD 844 slows growth of RAS mutant tumors and human epidermal growth factor receptor 2 (HER2)-overexpressing tumors, without causing hyperglycemia. Exhibits synergistic effects with Sotorasib (Cat. No. 7713) and Adagrasib (Cat. No. 7488). VVD 844 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 5-((1R,2S,5S)-2-(2-Chloro-3-fluorophenyl)-3-azabicyclo[3.1.0]hexan-3-yl)-N-((R,E)-4-(methylsulfonyl)but-3-en-2-yl)pyrazine-2-carboxamide |
| CAS | 3080226-63-4 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | GTPase Inhibitors |
| Molecular Formula | C21H22ClFN4O3S |
| Purity | 98% |