missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 6019650
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
VU 6019650 is a potent and selective M5 receptor antagonist (hM5 IC50 = 36 nM). Exhibits >100-fold selectivity over human and rat M1-4. In vivo, VU 6019650 inhibits the reward effects of oxycodone and reduces oxycodone self-administration in rats. VU 6019650 is blood brain barrier penetrant.
Specifications
Specifications
| Chemical Name or Material | (3R,4S)-rel-1-[(2,3-Dihydro-5-benzofuranyl)sulfonyl]-3-fluoro-4-[[(1-methyl-1H-imidazol-2-yl)thio]methyl]piperidine |
| CAS | 2926782-31-0 |
| Quantity | 10 mg |
| Target | Muscarinic Acetylcholine Receptor M5/CHRM5 |
| Molecular Formula | C18H22FN3O3S2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction