missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 6019650

Product Code. 30040318
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30040318

Brand: Tocris Bioscience™ 8055/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

M5 receptor antagonist

VU 6019650 is a potent and selective M5 receptor antagonist (hM5 IC50 = 36 nM). Exhibits >100-fold selectivity over human and rat M1-4. In vivo, VU 6019650 inhibits the reward effects of oxycodone and reduces oxycodone self-administration in rats. VU 6019650 is blood brain barrier penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,4S)-rel-1-[(2,3-Dihydro-5-benzofuranyl)sulfonyl]-3-fluoro-4-[[(1-methyl-1H-imidazol-2-yl)thio]methyl]piperidine
CAS 2926782-31-0
Quantity 50 mg
Target Muscarinic Acetylcholine Receptor M5/CHRM5
Molecular Formula C18H22FN3O3S2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.