missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0424465
GREENER_CHOICE

Product Code. 18766126
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18766126

Brand: Tocris Bioscience™ 6895/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent mGlu5 positive allosteric modulator and agonist; binds allosteric site with high affinity

VU 0424465 is a potent mGlu5 positive allosteric modulator (EC50 = 1.5 nM) and agonist (EC50 = 171 nM; maximum effect is 65% that of glutamate). Binds allosteric site with high affinity (Ki = 11.8 nM). Increases maximum response to glutamate by 30%. Exhibits bias towards signaling via IP1 and ERK1/2 over iCa2+ in HEK293 and neuronal cells, and activates Gs in HEK 293 cells. Induces epileptiform activity in CA3 hippocampal neurons in vitro and convulsions in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[2-(2-(3-Fluorophenyl)ethynyl]-N-[(1R)-2-hydroxy-1,2-dimethylpropyl]-2-pyridinecarboxamide
CAS 1428630-85-6
Quantity 10 mg
Molecular Formula C19H19FN2O2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.