missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0255035
GREENER_CHOICE

Product Code. p-7116043
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18779656

Brand: Tocris Bioscience™ 3727/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly selective M1 antagonist

VU 0255035 is a highly selective muscarinic M1 antagonist (Ki = 14.87 nM). Targets the M1 orthosteric site with greater than 75-fold selectivity over the other receptor subtypes. Reduces pilocarpine-induced seizures in mice. Brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-Oxo-3-[4-(4-pyridinyl)-1-piperazinyl]propyl]-2,1,3-benzothiadiazole-4-sulfonamide
CAS 1135243-19-4
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C18H20N6O3S2
Formula Weight Observed MW: 432.52
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.