Learn More
Heidolph™ Multi Reax Vortex Shaker
Product Code. 10627952 Shop All Heidolph Instruments Products
Change view
Click to view available options
Orbit:
3 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
10627952 3 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10627952 Supplier Heidolph™ Supplier No. 5451000000
  • ONLINE EXCLUSIVE PRICE
    1815.68€
    1643.00€ / Pack of 1
    Save 172.68€
    9% Off
View all promotions
Estimated Shipment: 02-12-2026
The Multi Reax Vortex Shaker includes an attachment that accommodates up to 26 vessels with various diameters. The 3 mm vibration orbit allows for excellent mixing results even in large and solid samples.
  • Unique shaker to accommodate up to 26 vessels with various diameters.
  • Includes attachment for 26 vessels with diameters from 10 to 16 mm and attachment for 12 vessels with diameters from 16 to 32 mm.
  • The 3 mm vibration orbit allows for excellent mixing results even in large and solid samples.
  • Set and continuously adjust the variable speeds of 150 to 2,000 rpm on the digital display.
  • Digital process timer allows for unattended operation and can be set from 1 to 999 minutes.
  • When the set time has elapsed, an alarm will sound and the operation stops.
TRUSTED_SUSTAINABILITY
Specifications
Orbit 3 mm
Load Bearing Capacity (Metric) 1.5 kg
Speed Range 150 to 2000 rpm
Electrical Requirements 230 V, 50 Hz
Temperature Range (Metric) 0°C to 50°C
Wattage 50 W
Load Bearing Capacity (English) 3.3 lb.
Certifications/Compliance DIN EN 60529 IP 30
Dimensions (L x W x H) 410 x 270 x 172 mm
Shipping Weight (English) 21.6 lb.
Shipping Weight (Metric) 9.8 kg
Weight (English) 21.6 lb.
Weight (Metric) 9.8 kg
Model Multi Reax
Display Digital
Includes 2 carousel attachments
No. of Positions 26
Timer Range 1 to 999 min.