Learn More
Socorex™ Acura™ 12-channel Micropipette Volumetric Module
Product Code. 10589482
Click to view available options
Volume (Metric):
10 μL
50 μL
350 μL
Unit Size:
Each
Product Code. 10589482
Brand: Socorex™ 800.12.050
Only null left
Add to basket
Add to basket
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ Acura™ 12-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your multi-channel micropipetting needs.
  • One of 27 available volumetric assemblies.
    • Increase versatility and save costs.
  • Disassembles and reassembles without tools.
  • Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
For Use With 12 Channel Pipette
Number of Channels 12
Product Line Acura
Type Micropipette Volumetric Module
Volume (Metric) 50 μL
For Use With (Application) 12 Channel Pipette
For Use With (Equipment) Acura™ Electro 926 Micropipette
Quantity 1 Each