Learn More
Witeg™ Volumetric Flask Class A with ST Blue Graduated
Product Code. 11837780
Click to view available options
Capacity (Metric):
20 mL
Unit Size:
Pack of 10
Product Code. 11837780
Brand: Witeg™ 3 664 00214 BL
Only null left
Add to basket
Add to basket
Borosilicate glass 3.3 volumetric flasks with ground joint neck and PE stopper, manufactured in accordance to DIN EN ISO 1042 and DE-M marked. Available in standard and special sizes, includes batch certificate.
  • Available with or without stopper
  • Borosilicate glass 3.3
  • Tolerance according to ASTM E 288
  • Batch certificate included
  • DE-M marked according to the German calibration law
  • Standard and special sizes available
Specifications
Calibration DE-M marked according to German calibration law
Capacity (English) 0.7 oz.
Closure Material Polyethylene
Capacity (Metric) 20 mL
Material Borosilicate Glass (3.3)
Neck Style Ground Joint
Socket Joint 14/23
Disposable Reusable
Graduated Yes
Sterility Non-sterile
Class A
Closure Type Stopper