Learn More
Afora™ Gasoline Volumetric Flask 10L
Click to view available options
Description:
Weight: 0.8kg
Unit Size:
Each
Description
- 10L
- Dark blue glass
Specifications
Specifications
| Color | Dark Blue |
| Description | Weight: 0.8kg |
| Capacity (Metric) | 10 L |
| Material | Glass |
| No. per Case | 1 Each |
| Bottom Shape | Flat |