missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[6-O-Acetyl-2,3,4-tris-O-(phenylmethyl)-Alpha-D-mannopyranosyl]-1-[(4-methylphenyl)sulfonyl]-1H-indole, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | 2-[6-O-Acetyl-2,3,4-tris-O-(phenylmethyl)-alpha-D-mannopyranosyl]-1-[(4-methylphenyl)sulfonyl]-1H-indole |
| CAS | 220339-91-3 |
| Molecular Formula | C44H43NO8S |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C44H43NO8S/c1-31-22-24-37(25-23-31)54(47,48)45-38-21-13-12-20-36(38)26-39(45)41-43(51-28-34-16-8-4-9-17-34)44(52-29-35-18-10-5-11-19-35)42(40(53-41)30-49-32(2)46)50-27-33-14-6-3-7-15-33/h3-26,40-44H,27-30H2,1-2H3/t40-,41-,42-,43-,44+/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@@H]([C@@H](OCc2ccccc2)[C@@H](OCc3ccccc3)[C@@H]1OCc4ccccc4)c5cc6ccccc6n5S(=O)(=O)c7ccc(C)cc7 |
| IUPAC Name | [(2R,3R,4R,5R,6R)-6-[1-(4-methylphenyl)sulfonylindol-2-yl]-3,4,5-tris(phenylmethoxy)oxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 745.88 |
| Formula Weight | 745.2709 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction