Learn More
Saint Gobain Life Sciences Natural Rubber Tubing
Artikelnummer. 11392084
Ansicht ändern
Click to view available options
Diameter (Metric) Inner:
8 mm
Packungsgröße:
25m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Diameter (Metric) Inner Packungsgröße
11392084 8 mm 25m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11392084 Lieferant Saint Gobain Life Sciences Lieferanten-Nr. 710110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Use this tubing for a variety of industrial, construction and chemical applications. Saint Gobain Life Sciences Natural Rubber Tubing provides an ideal combination of chemical resistance and pump life.
  • Natural Rubber Tubing
  • For vacuum applications
TRUSTED_SUSTAINABILITY
Spezifikation
Certifications/Compliance FDA compliant, Food WCR
Diameter (Metric) Inner 8 mm
Material Natural Rubber
Diameter (Metric) Outer 18 mm
Overall Length (Metric) 25 m
Thickness (Metric) 5 mm
For Use With (Application) General laboratory use
Min. Operating Temperature (Metric) -25°C
Color Red
Flexibility Flexible
Max. Operating Temperature (Metric) 70°C
Mehr anzeigen Weniger anzeigen