Learn More
Vitamin K3-d8, TRC
Specifications
Specifications
| Chemical Name or Material | Vitamin K3-d8 |
| CAS | 478171-80-1 |
| Molecular Formula | C11 2H8 O2 |
| Quantity | 10 mg |
| Synonym | 1,4-Naphthalenedione-2,5,6,7,8-d5','3-(methyl-d3)- (9CI','ACI), 3-(Methyl-d3)-1,4-naphthalenedione-2,5,6,7,8-d5 (ACI), Menadione-d8, Menadione-D8, Vitamin K3-d8, 2-Methyl-1,4-naphthoquinone-d8 |
| InChI Formula | InChI=1S/C11H8O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D |
| SMILES | [2H]C1=C(C(=O)c2c([2H])c([2H])c([2H])c([2H])c2C1=O)C([2H])([2H])[2H] |
| IUPAC Name | 2,5,6,7,8-pentadeuterio-3-(trideuteriomethyl)naphthalene-1,4-dione |
| Molecular Weight (g/mol) | 180.23 |
| Formula Weight | 180.1026 |