missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin K1-d7, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Vitamin K1-D7 |
| CAS | 1373049-34-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C31H39D7O2 |
| Quantity | 1 mg |
| Synonym | 1,4-Naphthalenedione-5,6,7,8-d4','2-(methyl-d3)-3-[(2E)-3,7,11,15-tetramethyl-2-hexadecen-1-yl]-, 2-(Methyl-d3)-3-[(2E)-3,7,11,15-tetramethyl-2-hexadecen-1-yl]-1,4-naphthalenedione-5,6,7,8-d4, trans-Phytomenadione-d7, trans-Phytonadione-d7, trans-Vitamin K1-d7, trans-Phylloquinone-d7, trans-Phytomenadione-D7 (5,6,7,8-D4','2-methyl-D3) |
| InChI Formula | InChI=1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/i6D3,7D,8D,18D,19D |
| SMILES | [2H]c1c([2H])c([2H])c2C(=O)C(=C(C\C=C(/C)\CCCC(C)CCCC(C)CCCC(C)C)C(=O)c2c1[2H])C([2H])([2H])[2H] |
| Molecular Weight (g/mol) | 457.74 |
| Formula Weight | 457.3937 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction