missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A palmitate, 1.7 M.I.U./g

Product Code. p-7076601
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15485289

Brand: Thermo Scientific Alfa Aesar J63022.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. | CAS: 79-81-2 | C36H60O2 | 524.874 g/mol

Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. Other applications include as pharmaceutical product which is used for manufacturing aqueous solutions using solubilizing agents such as cremophor. It is also used in the fortification of fatty foods such as margarine, spreads, oils, milk and dairy products. It is used in cosmetics for the preparation of creams and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-81-2
Molecular Formula C36H60O2
Molecular Weight (g/mol) 524.874
MDL Number MFCD00019414
InChI Key VYGQUTWHTHXGQB-FFHKNEKCSA-N
Synonym retinyl palmitate, vitamin a palmitate, retinol palmitate, retinol, hexadecanoate, arovit, all-trans-retinyl palmitate, optovit-a, retinyl hexadecanoate, aquapalm, dispatabs tabs
PubChem CID 5280531
ChEBI CHEBI:17616
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC=C(C)C=CC=C(C)C=CC1=C(CCCC1(C)C)C

Specifications

Melting Point 28°C to 29°C
Packaging Glass bottle
Quantity 25 g
Beilstein 1917366
Solubility Information Soluble in chloroform,ether,and vegetable oils. Insoluble in water.
Formula Weight 524.87
Physical Form Low Melting Solid
Chemical Name or Material Vitamin A palmitate
compliance-icons
Product Identifier
  • Vitamin A Palmitate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Retinol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.