Learn More
VITLAB™ Vital Instrument Tray
Click to view available options
Height (Metric):
17 mm
Unit Size:
Pack of 5
Specifications
Specifications
| Dimensions (L x W x H) | 268 x 208 x 17 mm |
| Color | White |
| Height (Metric) | 17 mm |
| Length (Metric) | 268 mm |
| Material | Melamine |
| Type | Instrument Tray |
| Width (Metric) | 208 mm |
| Quantity | 5/Pk. |
| Shape | Flat |