Specifications
| Detection Method | Chemiluminescence |
| Includes | Goat Anti-Rabbit IgG, HRP conjugate, Visualizer™ Detection Reagent B, Visualizer™ Detection Reagent A |
| Quantity | 20 Blots |
| Packaging | 20 Blots |
| For Use With (Application) | For Western Blotting, Dot/Slot Blotting |
| Host Species | Rabbit |
| Product Type | Visualizer Western Blot Detection Kit |