Learn More
VistaLab™ Technologies Pipette Tips: Non-Sterile
For use with Ovation and MLA Brand pipetters
Brand:  VistaLab™ Technologies 9022
Additional Details : Weight : 0.86180kg
 View more versions of this product
Product Code. 16465124
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Non-Sterile tips packaged in VistaRak, VistaStak, Stacked Rack, Bulk
  • VistaRak: 5 racks, 192 tips/rack, VistaStak: 3 or 5 Layers of 192 tips, Stacked Rack: 3 or 5 layers of 200 tips, Bulk: 750-1000 tips in a plastic bag or containers
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
Specifications
250 μL
Non-sterile
Blue
Stacked Dense Rack
6 in.
Straight
200 Tips/Layer
No
Non-Filtered
Clear
Not Autoclavable
2.2 in.
Rack
Stacked Rack, 5 layer
MLA, Ovation