Learn More
Macherey-Nagel™ Visocolor™ ECO Test Kit
Allows determination of even low limiting values with accuracy. Macherey-Nagel™ Visocolor™ ECO Test Kits is a group of colorimetric and titrimetric test kits that use precisely dosed single reagents. The results are evaluated visually with high-quality color comparison cards.
Brand:  Macherey-Nagel™ 931016
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10054022
  • 53.90€ / 150 tests
Estimated Shipment: 28-08-2024
Description
Visocolor™ ECO colorimetric and titrimetric test kits achieve high sensitivity and accuracy by using single reagents that can be dosed precisely and by the ability to compensate for the turbidity and color of water samples. The results are evaluated visually with color comparison cards adjusted to the original colors of standard solutions or photometrically with the PF-3 and PF-12Plus photometers for quantitative evaluation.
Easy
  • Chemical analysis without further accessories
  • No extensive training necessary
  • Color-coded reagents with clear dosing instructions
  • Easy to understand instruction manual
Safe
  • Pictogram test instructions
  • Reaction basis according to international standards
  • Compensation of turbidity and color
Unique
  • High quality test kits
  • Business-prized refill packs available
  • Ecologically friendly disposal of used reagents
  • Delivered in cardboard box with plastic inlay
Specifications
Specifications
DPD
PF-12 Plus, PF-3
150 Determinations
0.1 to 2.0mg/L Cl2
ECO Aluminum Colorimetric Test Kit
Free Chlorine determination
Reagents, Color comparator, Measuring spoon/glass, Syringe, Instructions in a cardboard box with plastic insert
1 Each
Colorimetric
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOR ECO free chlorine 2
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contains : N,N-diethyl-1,4-phenylene diammonium sulphate, sulphuric acid