Learn More
Thermo Scientific™ VISIONlite™ Spectrophotometer Software
Product Code. 17778418 Shop All Thermo Scientific Products
Click to view available options
Type:
6-user Software License
VISIONlite 5 Software
VISIONlite 5 Software + Cable
VISIONlite ColorCalc Software
Wine Analysis Software
Unit Size:
Each
Product Code. 17778418
Brand: Thermo Scientific™ 869191700
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ ColorCalc software is a powerful stand-alone software package that adds the ability to perform numerous color calculations for a variety of applications. The software records the required transmission or reflectance spectrum and automatically performs calculations using the recorded data.
Thermo Scientific™ VISIONlite™ EnzLab software offers predefined methods and operation procedures for all standard enzymatic test kits including Citric Acid, Lactic Acid, D-Glucose, D-Fructose, Ethanol, Lactose and many more. EnzLab automates and facilitates enzymatic determinations, ensures correct calculations, and provides appropriate analytical reports.
Thermo Scientific™ VISIONlite™ Wine Analysis software allows users to perform an extensive list of predefined photometric, colorimetric and enzymatic analytical procedures common in wine and juice analysis. This PC software features a simple user interface, reliable data recording, and error-free evaluation of results within comprehensive reports.
VISIONlite Software enhancements include:
Scan:
  • Peak pick to identify and label wavelength peaks or valleys
  • Import, save or export data in native or CSV format for easy archiving and reporting
  • Auto scaling of data plus simple zoom controls for detailed examination of spectra 
Rate:  For SPECTRONIC 200 and GENESYS 30, measure multi-cell kinetics in parallel or serial mode
  • Automatic enzyme activity calculation
  • Automatic data storage plus import or export your data in CSV format
  • Measure up to 100 samples in a single data set 
Fixed:
  • Measure at one or multiple wavelengths
  • Apply factors and equations to your data to generate results directly
  • Addition, subtraction, ratio and baseline correction options
  • Run standard bio-tests using the preprogrammed protein and DNA methods
Quant: More standards and non-linear curve fitting options than can be accomplished under local control on Thermo Scientific SPECTRONIC 200 and GENESYS 30 Spectrophotometers
TRUSTED_SUSTAINABILITY
Specifications
Type Wine Analysis Software
For Use With (Equipment) Thermo Scientific GENESYS 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers, SPECTRONIC 200 and GENESYS 30
Includes DVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean.
Required Accessories Interface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers.