Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Controller
Shop All Schott Products
Click to view available options
Type:
VisiLED Standard Controller MC1100
Unit Size:
Each
Description
- The compact MC1100 allows the settings of various illumination parameters including light intensity, segment modes and change of illumination direction while the MC1500 offers this for two light heads.
- Extra-fine smooth dimming.
- MC1500 offers an external trigger to start strobe function frequencies netween 15-5 KHz.
- MC1500 offers optional PC control via RS232 and footswitch control.
- Advanced setting options for optimum contrast.
- Ergonomics and blindfold operation.
- MC1500 offers control of two channels.
- Save up to four complex illumination scenarios with MC1500.
Specifications
Specifications
| Type | VisiLED Standard Controller MC1100 |
| Certifications/Compliance | CE, UKCA, EMC-class B |
| Dimensions (L x W x H) | 90 x 152 x 25 mm |
| For Use With (Application) | Stereo Microscopy Illumination |
| For Use With (Equipment) | VisiLED Ringlights or Transmitted Light Stage |
| Includes | DC In, 1 Lighthead Connector, ESD plug |
| Lighting Type | LED |
Product Content Correction
Product Title
Spot an opportunity for improvement?Condividi una correzione di contenuto