Learn More
A&D Instruments Viscometer Sine-wave 0.3 - 1000 mPa.s
GREENER_CHOICE
Brand:  A&D Instruments SV-1A
Viscometer Sine-wave 0.3 - 1000 mPa.s
Product Code. 16621292
  • ONLINE EXCLUSIVE PRICE
    10211.25€
    9482.14€ / Each
    Save 729.11€
    7% Off
Only null left
Estimated Shipment: 10-04-2026