Learn More
Heidolph™ VISCO JET™ Impeller
Shop All Heidolph Instruments Products
Click to view available options
For Use With (Equipment):
170 - 300 mm Vessel
80 - 150 mm Vessel
Speed:
120 - 500 rpm
200 to 800 rpm
Material:
POM
Stainless Steel (V4A)
Unit Size:
Each
Description
- Useful for applications that involve viscous media such as pastes and gels that need a reliable stirring process without air entry
- Unique blade shape triggers an intrinsically dynamic flow, which shortens processing time with significantly better mixing results compared to conventional impellers
Specifications
Specifications
| Diameter (Metric) | 10 mm |
| Diameter (Metric) Shaft | 10 mm |
| Height (English) Blade | 60 mm |
| For Use With (Equipment) | 80 - 150 mm Vessel |
| Length (Metric) | 500 mm |
| Speed | 200 to 800 rpm |
| Dimensions (L x W x H) | 500 x 60 x 10 mm |
| Stirring Range | 50 to 150 mm |
| Material | Stainless Steel (V4A) |