Learn More
Heidolph™ VISCO JET™ Impeller
Product Code. 10000962 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
170 - 300 mm Vessel
80 - 150 mm Vessel
Speed:
120 - 500 rpm
200 to 800 rpm
Material:
POM
Stainless Steel (V4A)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Speed Material Unit Size
10000962 170 - 300 mm Vessel 120 - 500 rpm POM Each
10356252 80 - 150 mm Vessel 200 to 800 rpm Stainless Steel (V4A) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10000962 Supplier Heidolph™ Supplier No. 5091612100
Estimated Shipment: 28-10-2026
Ideal for mixing viscous media
  • Useful for applications that involve viscous media such as pastes and gels that need a reliable stirring process without air entry
  • Unique blade shape triggers an intrinsically dynamic flow, which shortens processing time with significantly better mixing results compared to conventional impellers
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 10 mm shaft
Diameter (Metric) Shaft 10 mm
For Use With (Equipment) 170 - 300 mm Vessel
Height (Metric) Blade 120 mm
Length (Metric) 500 mm
Speed 120 - 500 rpm
Weight (Metric) 0.30 kg
Material POM