Learn More
Thermo Scientific™ Violet Red Bile Glucose Agar (ISO)
Product Code. 15795298
Change view
Click to view available options
Fill Weight:
198 to 202 g
99.0-101.0g
Format:
125 mL sirop bottle
250 mL Sirop Bottle
Unit Size:
2 L
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format Unit Size
15795298 198 to 202 g 250 mL Sirop Bottle 2 L
15829662 99.0-101.0g 125 mL sirop bottle Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15795298 Supplier Thermo Scientific™ Supplier No. BO1329R
  • ONLINE EXCLUSIVE PRICE
    60.88€
    40.64€ / 2 L
    Save 20.24€
    33% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Edge
Add to basket
Use this agar for the selective and differential isolation of Enterobacteriaceae
Violet Red Bile Glucose Agar (ISO) is a selective differential medium for the isolation of Enterobacteriaceae
TRUSTED_SUSTAINABILITY
Specifications
Color Dark Purple
Content And Storage 2° to 25°C
Format 250 mL Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 198 to 202 g
Packaging Type 10 Sirop Bottles with screw cap closures in a box
Quantity 10/Pk.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.