Learn More
Brady™ Vinyl with Polyester Overlaminate Durable Marking Tape
Product Code. 11711625
Click to view available options
Color:
Black, White
Black, Yellow
Magenta, Yellow
Red, White
Dimensions (L x W):
16.5 m x 25 mm
16.5 m x 38 mm
16.5 m x 50 mm
16.5 m x 75 mm
Unit Size:
1 roll
Product Code. 11711625
Brand: Brady™ 055312
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Dimensions (L x W) 16.5 m x 38 mm
Finish High Gloss
For Use With (Application) Safety marking of obstructions, low hanging objects, pillars, passageways, aisles, and any other potentially hazardous areas or objects
Length (Metric) 16.5 m
Packaging Quantity 1 Roll
Temperature Range (Metric) -40°C to +80°C
Thickness 0.037 mm
Width (Metric) 38 mm
Adhesive Type Rubber-based Pressure-Sensitive
Color Magenta, Yellow
Material Vinyl
Product Type Durable Marking Tape
Type Marking Tape