Learn More
Chemsplash Versa Vinyl Disposable Gloves Powder Free, Style 4050
GREENER_CHOICE
Product Code. 30058362
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 2000
Product Code. 30058362
Brand: Chemsplash 4050.300.006
Only null left
Add to basket
Add to basket
Strong and durable single-use vinyl gloves for use in the medical, food, or cleaning industry. Packaged in a box of 200.
  • Medical Grade
  • Food Contact Approved
  • Ambidextrous
  • Rolled Cuff and Latex Free
  • PFAS Free
  • Touchscreen Compatible
  • Thickness: 3.1 mil, Weight: 4.5 g
  • AQL: 1.5
  • CE and UKCA Approved
  • Box of 200
  • Suitable for single use in the medical, food, or cleaning industry.
  • Vinyl material is strong, durable.
  • Ideal for BRC approved work areas and the Pharmaceutical Industry.
  • roduct does not contain PFAS, RoHS, POP, TSCA, REACH, or any Conflict Materials.
Specifications
Certifications/Compliance (Europe) CAT III, EN ISO: 374-4:2019, EN ISO: 21420:2020, EN ISO: 374-1:2016+A1:2018 Type B, EN ISO: 374-5:2016
Accelerator Free No
Applications Healthcare, Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Smooth
Finger Thickness (English) 3.1 mil
Finger Thickness (Metric) 0.078 mm
Latex Free Yes
Material Vinyl
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.078 mm
Powdered Powder Free
Product Type Vinyl Disposable Gloves
Size Large
Sterility Non-sterile
Thickness (English) 3.1 mil
Thickness (Metric) 0.078 mm
Glove Cuff Style Rolled