Learn More
Brady™ Porta-Pack™ Vinyl Cloth Wire Marker Book
Product Code. 10576103 Shop All Brady Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10576103 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10576103 Supplier Brady™ Supplier No. 031203
  • ONLINE EXCLUSIVE PRICE
    46.00€
    38.93€ / Pack of 1
    Save 7.07€
    15% Off
Estimated Shipment: 29-12-2026
This B-500 repositionable Porta-Pack™ Vinyl Cloth Wire Marker Book is a convenient way to ensure the wire markers you need are always on hand.
  • Choose from 15 different books featuring numbers, letters, special symbols and NEMA colors.
  • Each book comes with 10 pages and includes a terminal marker with each wire marker.
  • Over 250 different refill packs are also available (sold separately).
Specifications
Certifications/Compliance ASTM D 1000, ASTM D 2979
Description Porta-Pack Wire Marker Books
Adhesive Type Repositionable Rubber
Color Black on White
Finish Matte
Format Label
Material Vinyl
Product Type Wire and Cable Markers
Sterility Non-sterile
Display Text Jan-45
For Use With (Application) Wire and cable labelling
Includes 10 markers of each legend
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Label/Pk.
Quantity 1 Ea.
Temperature Range (English) -40°F to +176°F
Temperature Range (Metric) -40°C to +80°C
Thickness (English) 0.0088 in.
Thickness (Metric) 0.224 mm
Dimensions (L x W) 39.7 x 5.5 mm
Length (English) 1.56 in.
Width (English) 0.22 in.
Width (Metric) 5.5 mm
Width (English) Printable Area 0.22 in.
Height (English) 1.56 in.
Height (Metric) 39.7 mm
Diameter (Metric) 12 mm
Alternate Part Number PWM-PK-3