Learn More
Vinyl Carbamate, Vinyl Carbamate, TRC
Product Code. 30135754
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135754 100 mg 100mg
30132581 25 mg 25mg
30011600 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135754 Supplier Toronto Research Chemicals Supplier No. TRCV375000100MG
 Restricted Item
Estimated Shipment: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C3 H5 N O2
Molecular Weight (g/mol) 87.08
IUPAC Name ethenyl carbamate
SMILES NC(=O)OC=C
Specifications
CAS 15805-73-9
Quantity 100 mg
InChI Formula InChI=1S/C3H5NO2/c1-2-6-3(4)5/h2H,1H2,(H2,4,5)
Formula Weight 87.032
Chemical Name or Material Vinyl Carbamate