Learn More
Nitritex™ BioClean Nano 4™ Non-sterile Nitrile Gloves, ISO Class 4
Product Code. 15208268
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15208268 X-Large Pack of 1000
15218268 2X-Large Pack of 1000
15268258 X-Small Pack of 1000
15278258 Small Pack of 1000
15288258 Medium Pack of 1000
15298258 Large Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15208268 Supplier Nitritex™ Supplier No. NAN4XL
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
With textured fingers to provide enhanced grip. Nitritex™ BioClean Nano 4™ Non-sterile Nitrile Gloves, ISO Class 4 are accelerator-free, latex-free, resistant to a wide range of chemicals and are suitable for use in electrically sensitive applications.
  • Resistant to a wide range of chemicals
  • Textured fingers for enhanced grip
  • Powder-, sulfur-, latex- and accelerator-free
  • Excellent antistatic properties
  • Ultra-low particle count: <800
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) ISO 9001:2015 and (EU)2016/425, EN420:2003 + A1:2009, EN374-1:20016, EN374-5:2016 and EN421:2010, EN455-2:2015, (EC)1935/2004 and (EC)2023/2006
Color White
Disposable Single-use
Exterior Finish Fully Textured
Grade Category 3 - PPE Directive 89/686/EEC
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Finger Style Textured
For Use With (Application) Electrically sensitivity applications
Glove Cuff Style Beaded
Quantity 100 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces)
Resistant To A broad range of chemicals
Series BioClean Nano 4
Length (English) 12 in.
Length (Metric) 30 cm
Width (Metric) 115 mm ± 5