missing translation for 'onlineSavingsMsg'
Learn More

Vincristine sulfate, 99.4%, MP Biomedicals™

Product Code. 11426111 Shop All MP Biomedical Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg

Product Code. 11426111

Brand: MP Biomedicals 0219068705

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 2068-78-2
Molecular Formula C46H58N4O14S
Molecular Weight (g/mol) 923.044
InChI Key AQTQHPDCURKLKT-DJOZZVTQSA-N
Synonym vincristine sulfate, methyl 9r,10s,11r,12r,19r-11-acetyloxy-12-ethyl-4-13s,15r,17s-17-ethyl-17-hydroxy-13-methoxycarbonyl-1,11-diazatetracyclo 13.3.1.0?, 1 2 .0?, 1 ? nonadeca-4 12 ,5,7,9-tetraen-13-yl-8-formyl-10-hydroxy-5-methoxy-8,16-diazapentacyclo 10.6.1.0 1 ,?.0 2 ,?.0
PubChem CID 126963557
SMILES CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O

Specifications

Color White
Assay Percent Range 0.994
Quantity 5 mg
Formula Weight 923
Physical Form Powder
Chemical Name or Material Vincristine sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.