Learn More
Thermo Scientific Chemicals Vincristine sulfate, 95%
Product Code. 10731481 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mg
Packaging:
Glass Bottle
Unit Size:
5mg
Product Code. 10731481
Brand: Thermo Scientific Chemicals 203440050
Request Bulk or Custom Format
CAS: 2068-78-2 | C46H56N4O10·H2SO4 | 923.03 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 2068-78-2
Melting Point >300°C
Color White to Yellow
Boiling Point 273°C to 281°C
Loss on Drying 12% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C46H56N4O10·H2SO4
MDL Number MFCD08706469
Quantity 5 mg
Merck Index 15, 10183
Synonym vincristine sulfate, methyl 9r,10s,11r,12r,19r-11-acetyloxy-12-ethyl-4-13s,15r,17s-17-ethyl-17-hydroxy-13-methoxycarbonyl-1,11-diazatetracyclo 13.3.1.0?, 1 2 .0?, 1 ? nonadeca-4 12 ,5,7,9-tetraen-13-yl-8-formyl-10-hydroxy-5-methoxy-8,16-diazapentacyclo 10.6.1.0 1 ,?.0 2 ,?.0
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,slightly soluble in alcohol,practically insoluble in ether
InChI Key AQTQHPDCURKLKT-DJOZZVTQSA-N
SMILES CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
Molecular Weight (g/mol) 923.03
PubChem CID 126963557
Formula Weight 923.03
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Vincristine sulfate, 95%