Learn More
BINDER™ Viewing Window and Interior Lighting
Product Code. p-7237272 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers ED 260
BINDER Chambers ED 56
BINDER Chambers ED 720
BINDER Chambers FD 23
BINDER Chambers FD 260
BINDER Chambers FD 56
BINDER Chambers FD 720
BINDER Chambers FED 260
BINDER Chambers FED 400
BINDER Chambers FED 56
Unit Size:
Each
Product Code. 16484049
Brand: BINDER™ 80121058
Only null left
Add to basket
Add to basket
This Viewing Window and Interior Lighting is designed for use with assorted BINDER™ Drying and Heating Chambers. Insulated viewing window and interior lighting allows observation without interfering with internal conditions.
  • Viewing window with triple insulating glass in door(s)
  • 15 or 30 W interior lighting, depending on the model
Specifications
Description Viewing window and interior lighting - 230 V option model, viewing window (triple insulating glass) in door, 240 x 240 mm, and 15 W interior
For Use With (Equipment) BINDER Chambers ED 56
Type Viewing Window with Interior Lighting
Insulation Triple Insulating Glass
Width (Metric) 240 mm
Height (Metric) 240 mm
Wattage 15 W