Learn More
BINDER™ Viewing Window and Interior Lighting
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers ED 260
BINDER Chambers ED 56
BINDER Chambers ED 720
BINDER Chambers FD 23
BINDER Chambers FD 260
BINDER Chambers FD 56
BINDER Chambers FD 720
BINDER Chambers FED 260
BINDER Chambers FED 400
BINDER Chambers FED 56
Unit Size:
Each
Description
- Viewing window with triple insulating glass in door(s)
- 15 or 30 W interior lighting, depending on the model
Specifications
Specifications
| Description | Viewing window and interior lighting - 230 V option model, viewing window (triple insulating glass) in door, 370 x 370 mm, and 15 W interior |
| For Use With (Equipment) | BINDER Chambers ED 260 |
| Type | Viewing Window with Interior Lighting |
| Insulation | Triple Insulating Glass |
| Width (Metric) | 370 mm |
| Height (Metric) | 370 mm |
| Wattage | 15 W |