Learn More
Prenylamine, TRC
Product Code. 30010725
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010725 50 mg 50mg
30004205 100 mg 100mg
30005913 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010725 Supplier Toronto Research Chemicals Supplier No. TRCP71280050MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 390-64-7
Molecular Formula C24 H27 N
Molecular Weight (g/mol) 329.48
Synonym Benzenepropanamine, N-(1-methyl-2-phenylethyl)-γ-phenyl-, Phenethylamine, N-(3,3-diphenylpropyl)-α-methyl- (6CI,8CI), N-(1-Methyl-2-phenylethyl)-γ-phenylbenzenepropanamine, B 436, DL-Prenylamine, Elecor, N-(3,3-Diphenylpropyl)-α-methylphenethylamine, N-(3,3-Diphenylpropyl)-α-methylphenylethylamine, Prenylamine, Prenylamine (vasodilator), SAN 13-194, Sandoz 13-194, Segontin
IUPAC Name 3,3-diphenyl-N-(1-phenylpropan-2-yl)propan-1-amine
SMILES CC(Cc1ccccc1)NCCC(c2ccccc2)c3ccccc3
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C24H27N/c1-20(19-21-11-5-2-6-12-21)25-18-17-24(22-13-7-3-8-14-22)23-15-9-4-10-16-23/h2-16,20,24-25H,17-19H2,1H3
Formula Weight 329.2143
Percent Purity >95%
Chemical Name or Material Prenylamine
Grade HPLC